Learn More
Chemglass Life Sciences Square Media Bottle, Sterile PETG, Graduated, 125 mL, 38-430 Tamperproof Cap, Pack of 24
SDP
Catalog No. NC3481250
Encompass
Sterile square PETG media bottle for storage and transport of culture media and laboratory solutions. USP Class VI PETG construction provides optical clarity and reduced permeability to gases; gamma sterilized and free of pyrogens, DNase, RNase, and particles. Tamperproof cap offers a visual seal indicator; not autoclavable.
  • Gamma sterilized.
  • Manufactured from USP Class VI PETG.
  • 125 mL capacity.
  • 38-430 cap/thread size.
  • Tamperproof polypropylene cap provides visible seal indication.
  • Pyrogen-, DNase-, RNase-, and particle-free.
  • Reduced permeability to CO2 and O2.
  • Not autoclavable.
  • Pack of 24 (P24) for tamperproof cap variant.
Catalog No. NC3481250 Supplier Chemglass Life Sciences Supplier No. CLS1168X0125
Sterile square PETG media bottle for storage and transport of culture media and laboratory solutions. USP Class VI PETG construction provides optical clarity and reduced permeability to gases; gamma sterilized and free of pyrogens, DNase, RNase, and particles. Tamperproof cap offers a visual seal indicator; not autoclavable.
  • Gamma sterilized.
  • Manufactured from USP Class VI PETG.
  • 125 mL capacity.
  • 38-430 cap/thread size.
  • Tamperproof polypropylene cap provides visible seal indication.
  • Pyrogen-, DNase-, RNase-, and particle-free.
  • Reduced permeability to CO2 and O2.
  • Not autoclavable.
  • Pack of 24 (P24) for tamperproof cap variant.