Learn More
Chemglass Life Sciences Chemglass CLS-1168-0250 single-use PETG media bottle assembly, 250 mL, 2-port closure, sterile, pack of 10
SDP
Catalog No. NC3479977
Encompass
Single-use 250 mL PETG media bottle assembly designed for sterile bioprocessing and media transfer. The assembly includes a two-port closure, vent filter, and V-notched dip tube to maximize drainability; materials are USP Class VI and animal derived component free, and units are gamma irradiated to SAL 10^-6.
  • 250 mL PETG bottle with molded graduations.
  • Two-port closure assembly for transfer and venting.
  • 30 mm, 0.22 μm vent filter.
  • V-notched dip tube extends to bottom for maximum drainability.
  • 24 in length of weldable TPE tubing (models with TPE tubing).
  • Stopper molded with strain relief to prevent kinking.
  • Gamma irradiated to Sterility Assurance Level (SAL) 10^-6 and double poly-bagged.
  • Manufactured, assembled, and packaged in a Class 7 cleanroom.
  • USP Class VI approved materials and animal derived component free (ADCF).
  • High-clarity, break-resistant design; not autoclavable.
Catalog No. NC3479977 Supplier Chemglass Life Sciences Supplier No. CLS1168X0250
Single-use 250 mL PETG media bottle assembly designed for sterile bioprocessing and media transfer. The assembly includes a two-port closure, vent filter, and V-notched dip tube to maximize drainability; materials are USP Class VI and animal derived component free, and units are gamma irradiated to SAL 10^-6.
  • 250 mL PETG bottle with molded graduations.
  • Two-port closure assembly for transfer and venting.
  • 30 mm, 0.22 μm vent filter.
  • V-notched dip tube extends to bottom for maximum drainability.
  • 24 in length of weldable TPE tubing (models with TPE tubing).
  • Stopper molded with strain relief to prevent kinking.
  • Gamma irradiated to Sterility Assurance Level (SAL) 10^-6 and double poly-bagged.
  • Manufactured, assembled, and packaged in a Class 7 cleanroom.
  • USP Class VI approved materials and animal derived component free (ADCF).
  • High-clarity, break-resistant design; not autoclavable.