Learn More
Chemglass Life Sciences Chemglass Media Bottle (Square), Sterile PETG, 500 mL, 38-430 Tamperproof Cap
SDP
Catalog No. NC3481668
Encompass
Sterile, square 500 mL PETG media bottle designed for laboratory media storage and handling. Gamma-irradiated and packaged under cleanroom conditions to minimize contamination risk.
  • Sterile, gamma-irradiated for single use.
  • 500 mL capacity.
  • Square shape for stable storage and efficient shelving.
  • PETG construction, USP Class VI approved.
  • 38-430 tamperproof polypropylene cap.
  • Non-pyrogenic, DNase- and RNase-free.
  • Double poly-bagged and packaged in a Class 7 cleanroom.
Catalog No. NC3481668 Supplier Chemglass Life Sciences Supplier No. CLS1168X0500
Sterile, square 500 mL PETG media bottle designed for laboratory media storage and handling. Gamma-irradiated and packaged under cleanroom conditions to minimize contamination risk.
  • Sterile, gamma-irradiated for single use.
  • 500 mL capacity.
  • Square shape for stable storage and efficient shelving.
  • PETG construction, USP Class VI approved.
  • 38-430 tamperproof polypropylene cap.
  • Non-pyrogenic, DNase- and RNase-free.
  • Double poly-bagged and packaged in a Class 7 cleanroom.