Learn More
Chemglass Life Sciences 1, 000mL Square Media Bottle, Graduated, GL-45
SDP
Catalog No. NC1736547
Encompass_Preferred
1,000mL Square Media Bottle, Graduated, GL-45. Square bottles are easier to handle, require less space (13-20%) on the shelf or in the autoclave and are ideal for mixing, sampling and storage. Features include: white enamel graduations, large marking spot and a glass ridge molded into the bottle to indicate full capacity. Made from Pyrex borosilicate glass for chemical and thermal resistance and includes a liner less one-piece autoclavable orange polypropylene plug seal cap with drip-free pouring ring. CAUTION: DO NOT have caps tightened immediately after autoclaving as the vacuum resulting from cooling can cause breakage. Not applicable for vacuum use.
Catalog No. NC1736547 Supplier Chemglass Life Sciences Supplier No. CG80921L
1,000mL Square Media Bottle, Graduated, GL-45. Square bottles are easier to handle, require less space (13-20%) on the shelf or in the autoclave and are ideal for mixing, sampling and storage. Features include: white enamel graduations, large marking spot and a glass ridge molded into the bottle to indicate full capacity. Made from Pyrex borosilicate glass for chemical and thermal resistance and includes a liner less one-piece autoclavable orange polypropylene plug seal cap with drip-free pouring ring. CAUTION: DO NOT have caps tightened immediately after autoclaving as the vacuum resulting from cooling can cause breakage. Not applicable for vacuum use.