Learn More
Hach Company Sq. Storage Bottle PE 60 mL
Supplier:  Hach Company 2086871
Encompass
Bottle-Storage-PE-Square-60 mL-12/pkHigh-density polyethylene bottle with leak-proof polypropylene cap Square design saves space while wide-mouth allows for easy fill and removal of contents
Catalog No. 50-003-04264