Learn More
Accuflow Systems Eagle Thermoplastics Square Weighing Dish, Polystyrene, 1-5/8 in, 100/pack
Catalog No. NC1751508
Encompass_Preferred
Eagle Thermoplastics square polystyrene weighing dishes provide an anti-static, shallow sample vessel designed for precise analytical weighing and routine sample handling. The square geometry and shallow depth make these dishes well suited for small balance pans, moisture analyzers, and rapid sample transfers.
  • Anti-static rubberized polystyrene reduces sample adhesion and static errors.
  • Square footprint, 1-5/8 in top internal dimension, fits small balance pans.
  • Shallow 5/16 in depth for easy transfer and quick drying.
  • Available in natural and blue, 100 per pack for routine laboratory use.
  • Disposable design simplifies cleanup and prevents cross-contamination.
Catalog No. NC1751508 Supplier Accuflow Systems Supplier No. WB158100
Eagle Thermoplastics square polystyrene weighing dishes provide an anti-static, shallow sample vessel designed for precise analytical weighing and routine sample handling. The square geometry and shallow depth make these dishes well suited for small balance pans, moisture analyzers, and rapid sample transfers.
  • Anti-static rubberized polystyrene reduces sample adhesion and static errors.
  • Square footprint, 1-5/8 in top internal dimension, fits small balance pans.
  • Shallow 5/16 in depth for easy transfer and quick drying.
  • Available in natural and blue, 100 per pack for routine laboratory use.
  • Disposable design simplifies cleanup and prevents cross-contamination.