Learn More
Medchemexpress LLC HY-14989 5mg Medchemexpress, SQ109 CAS:502487-67-4 Purity:>98%
SDP
Catalog No. 502030574
Encompass_Preferred
Medchemexpress, HY-14989 5mg SQ109 CAS:502487-67-4 SQ109 is a potent inhibitor of the trypomastigote form of the parasite, with IC50 for cell killing of 50±8 nM. SQ109, targets MmpL3, is an antitubercular agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0574 Supplier Medchemexpress LLC Supplier No. HY149895MG
Medchemexpress, HY-14989 5mg SQ109 CAS:502487-67-4 SQ109 is a potent inhibitor of the trypomastigote form of the parasite, with IC50 for cell killing of 50±8 nM. SQ109, targets MmpL3, is an antitubercular agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.