Learn More
Heidolph™ SQ13 Screwthread Adapters
SDP
Catalog No. 05404115 Shop All Heidolph North America Products
Change view
Click to view available options
Type:
B29
Catalog No. Type
05-404-115 B29
1 options
Catalog No. 05-404-115 Supplier Heidolph™ Supplier No. 015890880
Add to Cart
Add to Cart
Heidolph™ SQ13 Screwthread Adapters are available in two different sizes
  • Adapter to SQ13 Screwthread
  • Available in two different types
Specifications
For Use With (Equipment) SQ13 Screwthread
For Use With (Application) Allows for Connection of Gas or Vacuum
Quantity 5/Pk.
Type B29
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov