Learn More
Medchemexpress LLC HY-100396 10mg , SQ22536 CAS:17318-31-9 Purity:>98%
SDP
Catalog No. 502259413
Encompass_Preferred
HY-100396 10mg SQ22536 CAS: 17318-31-9 SQ22536 is an effective adenylate cyclase (AC) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9413 Supplier Medchemexpress LLC Supplier No. HY10039610MG
HY-100396 10mg SQ22536 CAS: 17318-31-9 SQ22536 is an effective adenylate cyclase (AC) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.