Learn More
TARGETMOL CHEMICALS INC SQ22536 25MG
SDP
Catalog No. 502581009
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SQ22536 (9-(tetrahydrofuran-2-yl)-9h-purin-6-amine) the adenosine analogue 9-(Tetrahydro-2-furyl)adenine inhibited adenylate cyclase activity of crude membrane preparations from catfish (Ictalurus melas) and rat isolated hepatocytes in a non-competitive manner. purity: 98%
Catalog No. 50-258-1009 Supplier TARGETMOL CHEMICALS INC Supplier No. T217225MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SQ22536 (9-(tetrahydrofuran-2-yl)-9h-purin-6-amine) the adenosine analogue 9-(Tetrahydro-2-furyl)adenine inhibited adenylate cyclase activity of crude membrane preparations from catfish (Ictalurus melas) and rat isolated hepatocytes in a non-competitive manner. purity: 98%