Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Standards Kit, Mass Spectrometry, 6 Components, System Assessment

Catalog No. 502390682 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Mass Spectrometry (SQD) standards kit is designed for system assessment. It contains six components in various solutions for Mass Spectrometry applications. Each kit is shipped with a Certificate of Analysis. Classified as hazardous material, it requires special handling and should be stored at a temperature of less than 4°C in a firmly sealed, corrosion-proof container.

  • Contains 6 components.
  • Designed for reversed phase separation.
  • Utilizes MS detection.
  • Liquid formulation.
  • For MS system type.
  • Shelf life of 12 months.
  • Ships under ambient conditions.
  • Reference type.
  • Application for system performance.
  • Product type is standards.
  • 1 unit per package.
  • Hazardous material requiring special freight handling and additional charges.
  • Store at a temperature of less than 4°C.

Catalog No. 50-239-0682 Supplier Waters Corp Supplier No. 700003093
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Mass Spectrometry (SQD) standards kit is designed for system assessment. It contains six components in various solutions for Mass Spectrometry applications. Each kit is shipped with a Certificate of Analysis. Classified as hazardous material, it requires special handling and should be stored at a temperature of less than 4°C in a firmly sealed, corrosion-proof container.

  • Contains 6 components.
  • Designed for reversed phase separation.
  • Utilizes MS detection.
  • Liquid formulation.
  • For MS system type.
  • Shelf life of 12 months.
  • Ships under ambient conditions.
  • Reference type.
  • Application for system performance.
  • Product type is standards.
  • 1 unit per package.
  • Hazardous material requiring special freight handling and additional charges.
  • Store at a temperature of less than 4°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.