Learn More
Waters Corp Performance Maintenance Kit, SQD/TQD/3100, Oil Free, Chemical Kit
Catalog No. 50465912 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This annual performance maintenance kit is designed for SQD/TQD/3100 oil-free mass spectrometers. It includes source and ESI probe components, along with a chemical kit necessary for performance testing to ensure optimal equipment performance.
  • Performance maintenance type
  • For MS system type
  • Annual replacement schedule
  • Includes source components
  • Includes ESI probe components
  • Includes chemical kit for performance testing
Catalog No. 50-465-912 Supplier Waters Corp Supplier No. 176002135
This annual performance maintenance kit is designed for SQD/TQD/3100 oil-free mass spectrometers. It includes source and ESI probe components, along with a chemical kit necessary for performance testing to ensure optimal equipment performance.
  • Performance maintenance type
  • For MS system type
  • Annual replacement schedule
  • Includes source components
  • Includes ESI probe components
  • Includes chemical kit for performance testing