Learn More
Waters Corp Waters Performance Maintenance Kit, SQD/TQD/3100, Rotary
Catalog No. 50791594 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters™ Performance Maintenance Kit for the SQD/TQD/3100 Mass Spectrometer contains all the Waters Quality Parts™ necessary to keep your instrument operating at peak performance.
  • Includes source components, ESI probe components, and vacuum pump components.
  • Designed for use with 3100 Mass Detector, ACQUITY SQ Detector, and ACQUITY TQ Detector instruments.
  • Replacement schedule: Annual.
  • Product type: Performance maintenance kits.
Catalog No. 50-791-594 Supplier Waters Corp Supplier No. 201000216
The Waters™ Performance Maintenance Kit for the SQD/TQD/3100 Mass Spectrometer contains all the Waters Quality Parts™ necessary to keep your instrument operating at peak performance.
  • Includes source components, ESI probe components, and vacuum pump components.
  • Designed for use with 3100 Mass Detector, ACQUITY SQ Detector, and ACQUITY TQ Detector instruments.
  • Replacement schedule: Annual.
  • Product type: Performance maintenance kits.