Learn More
Aviva Systems Biology Corp SQLE Antibody - C-terminal region (ARP42101_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42101_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SQLE. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human SQLE. Alternate names: FLJ30795ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-58107