Learn More
MilliporeSigma SQPAK™ Quanta Cartridge
Catalog No. SQPAKQUAM1 Shop All MilliporeSigma Products
Change view
Click to view available options
Purification Method:
Ion Exchange
Catalog No. Purification Method
SQPAKQUAM1 Ion Exchange
1 options
Catalog No. SQPAKQUAM1 Supplier MilliporeSigma Supplier No. SQPAKQUAM1
Get an Extra Lab Water Cartridge with Purchase
View all promotions
Add to Cart
Add to Cart
Produces ultrapure (Type 1) water for use on the ultrapure dispensing module of Milli-Q™ SQ 2Series systems
  • Removes ions from pure water.
  • Produce Ultrapure (Type 1) water at 18.2 MΩ·cm resistivity.
  • Easy replacement when exhausted thanks to the Twist and Lock mechanism.
  • For use on the ultrapure dispensing module of Milli-Q™ SQ 2Series configurations, including Milli-Q™ SQ 200/200P/240C/240CV systems
TRUSTED_SUSTAINABILITY
Specifications
Purification Method Ion Exchange
Resin Type Ion Exchange Resin