Learn More
MilliporeSigma SQPAK™ RO Cartridge
Catalog No. SQPAKPR0M1 Shop All MilliporeSigma Products
Change view
Click to view available options
For Use With (Equipment):
Milli-Q SQ 2 Series
Catalog No. For Use With (Equipment)
SQPAKPR0M1 Milli-Q SQ 2 Series
1 options
Catalog No. SQPAKPR0M1 Supplier MilliporeSigma Supplier No. SQPAKPR0M1
Add to Cart
Add to Cart
Removes organics, ions, particulates up to 99% from tap water feeding Milli-Q™ SQ 2Series RO production stations
  • Removes organics, ions, bacteria and particulates up to 99% from tap water feed.
  • Produce pure reverse osmosis (RO) water at 42 L/hr.
  • Easy replacement when exhausted thanks to the Twist and Lock mechanism.
  • For use on the pure RO production stations of Milli-Q™ SQ 2Series configurations, including Milli-Q™ SR 240/240L/240XL and Milli-Q™ SQ 240C/240CV systems
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Milli-Q SQ 2 Series
Type Cartridge, RO