Learn More
Electron Microscopy Sciences Athene 200 mesh Square Center Mark Grid, Au
SDP
Catalog No. 501926665
Encompass_Preferred
Athene grids are renowned for the highest quality standards, exceptionally refined grid bars, and good handling characteristics. 3.05 mm diameter.
Grid: Gold
Catalog No. 50-192-6665 Supplier Electron Microscopy Sciences Supplier No. AC200AU
Athene grids are renowned for the highest quality standards, exceptionally refined grid bars, and good handling characteristics. 3.05 mm diameter.
Grid: Gold