Learn More
Aviva Systems Biology Corp SQOR Antibody - middle region (ARP62719_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62719_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SQRDL. It was validated on Western Blot. . Alternate names: CGI-44ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-287-754