Learn More
Aviva Systems Biology Corp SQOR Antibody - C-terminal region (ARP62718_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62718_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Sqrdl. It was validated on Western Blot. . Alternate names: 0610039J17Rik, 4930557M22RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-287-753