Learn More
Aviva Systems Biology Corp SQSTM1 Antibody - middle region (ARP54262_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54262_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SQSTM1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SQSTM1. Alternate names: A170, OSIL, PDB3, ZIP3, p60, p62, p62BApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-61149