Learn More
Medchemexpress LLC Squalane-d62 | 16514-83-3 | 100 MG
SDP
Catalog No. 5000417642
Encompass_Preferred
Squalane-d62 is the deuterium-labeled version of Squalane, a saturated derivative of Squalene found in certain fish and vegetable oils. It exhibits anticancer, antioxidant, skin hydrating, and emollient activities.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-17642 Supplier Medchemexpress LLC Supplier No. HYN7113S100MG
Squalane-d62 is the deuterium-labeled version of Squalane, a saturated derivative of Squalene found in certain fish and vegetable oils. It exhibits anticancer, antioxidant, skin hydrating, and emollient activities.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS