Learn More
Medchemexpress LLC Squalane-d62 | 16514-83-3 | 25 MG
SDP
Catalog No. 5000421522
Encompass_Preferred
Squalane-d62 is a deuterium-labeled form of Squalane, a saturated derivative of Squalene found in certain fish and vegetable oils. It exhibits anticancer, antioxidant, skin hydrating, and emollient activities. This product is intended for research use only. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules primarily as tracers for quantitation during drug development, and can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable under recommended storage conditions.
Catalog No. 50-004-21522 Supplier Medchemexpress LLC Supplier No. 25MGHYN7113S
Squalane-d62 is a deuterium-labeled form of Squalane, a saturated derivative of Squalene found in certain fish and vegetable oils. It exhibits anticancer, antioxidant, skin hydrating, and emollient activities. This product is intended for research use only. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules primarily as tracers for quantitation during drug development, and can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable under recommended storage conditions.