Learn More
Medchemexpress LLC Squalane-d62 | 16514-83-3 | 5 MG
SDP
Catalog No. 5000418289
Encompass_Preferred
Squalane-d62 is the deuterium labeled Squalane, a saturated derivative of Squalene found in certain fish and vegetable oils. It exhibits anticancer, antioxidant, skin hydrating, and emollient activities. Deuteration, incorporating stable heavy isotopes of hydrogen, carbon, and other elements, has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules as tracers for quantitation during drug development.
Catalog No. 50-004-18289 Supplier Medchemexpress LLC Supplier No. HYN7113S5MG
Squalane-d62 is the deuterium labeled Squalane, a saturated derivative of Squalene found in certain fish and vegetable oils. It exhibits anticancer, antioxidant, skin hydrating, and emollient activities. Deuteration, incorporating stable heavy isotopes of hydrogen, carbon, and other elements, has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules as tracers for quantitation during drug development.