Learn More
Medchemexpress LLC Squalane-d62 | 16514-83-3 | 50 MG
SDP
Catalog No. 5000419763
Encompass_Preferred
Squalane-d62 is a deuterium-labeled Squalane, a saturated derivative of Squalene found in certain fish oils and vegetable oils. It demonstrates anticancer, antioxidant, skin hydrating, and emollient activities. Deuterated compounds, like Squalane-d62, incorporate stable heavy isotopes and are primarily used as tracers for quantitation during drug development, impacting pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-19763 Supplier Medchemexpress LLC Supplier No. HYN7113S50MG
Squalane-d62 is a deuterium-labeled Squalane, a saturated derivative of Squalene found in certain fish oils and vegetable oils. It demonstrates anticancer, antioxidant, skin hydrating, and emollient activities. Deuterated compounds, like Squalane-d62, incorporate stable heavy isotopes and are primarily used as tracers for quantitation during drug development, impacting pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS