Learn More
Medchemexpress LLC Squalene | 111-02-4 | 410.72 | 100 MG
SDP
Catalog No. 5000415822
Encompass_Preferred
Squalene is an intermediate product in the synthesis of cholesterol. It exhibits several pharmacological properties, including hypolipidemic, hepatoprotective, antiatherosclerotic, cardioprotective, antioxidant, and antitumour activity.
  • Intermediate product in cholesterol synthesis
  • Shows hypolipidemic properties
  • Exhibits hepatoprotective effects
  • Possesses antiatherosclerotic activity
  • Provides cardioprotective benefits
  • Acts as an antioxidant
  • Demonstrates antitumour activity
  • Purity: 98.0%
Catalog No. 50-004-15822 Supplier Medchemexpress LLC Supplier No. HYN1214100MG
Squalene is an intermediate product in the synthesis of cholesterol. It exhibits several pharmacological properties, including hypolipidemic, hepatoprotective, antiatherosclerotic, cardioprotective, antioxidant, and antitumour activity.
  • Intermediate product in cholesterol synthesis
  • Shows hypolipidemic properties
  • Exhibits hepatoprotective effects
  • Possesses antiatherosclerotic activity
  • Provides cardioprotective benefits
  • Acts as an antioxidant
  • Demonstrates antitumour activity
  • Purity: 98.0%