Learn More
Selleck Chemical LLC Squalene S4862-25mg
SDP
Catalog No. 503111392
Encompass_Preferred
Squalene (Spinacene Supraene trans-Squalene) a naturally occurring substance found in plants animals and humans is a component of some adjuvants that is added to vaccines to enhance the immune response
Catalog No. 50-311-1392 Supplier Selleck Chemical LLC Supplier No. S486225MG
Squalene (Spinacene Supraene trans-Squalene) a naturally occurring substance found in plants animals and humans is a component of some adjuvants that is added to vaccines to enhance the immune response