Learn More
Medchemexpress LLC Squalene-d6 | 1875101-86-2 | 416.75 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN1214S5MG
Encompass_Preferred
Squalene-d6 (Super Squalene-d6) is a deuterium labeled Squalene. Squalene (Super Squalene) is an intermediate product in the synthesis of cholesterol, and shows several pharmacological properties such as hypolipidemic, hepatoprotective, antiatherosclerotic, cardioprotective, antioxidant, and antitumour activity.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-15499