Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Squalene, MP Biomedicals™

Catalog No. mp210294891 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210294891 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210294891 Supplier MP Biomedicals, Inc Supplier No. 0210294891
Only null left
Add to Cart
Add to Cart

Functions as the biochemical precursor to the whole family of steroids. Squalene, MP Biomedicals is an isoprenoid compound that is structurally similar to beta-carotene and is an intermediate metabolite in the synthesis of cholesterol. Squalene is not ver

  • Functions in the skin as a quencher of singlet oxygen, protecting human skin surface from lipid peroxidation due to exposure to UV and other sources of ionizing radiation
  • Supplementation in mice has resulted in marked increases in cellular and non-specific immune functions in a dose-dependent manner
  • May act as a “sink” for highly lipophilic xenobiotics; since it is a nonpolar substance, it has a higher affinity for un-ionized drugs
  • Supplementation can reduce cholesterol and triglyceride levels; it might be a useful addition to potentiate the effects of some cholesterol-lowering drugs
  • The primary therapeutic use of squalene currently is as an adjunctive therapy in a variety of cancers
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-02-4
Molecular Formula C30H50
Molecular Weight (g/mol) 410.73
InChI Key YYGNTYWPHWGJRM-AAJYLUCBSA-N
Synonym squalene, trans-squalene, spinacene, supraene, all-trans-squalene, 2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene, e,e,e,e-squalene, unii-7qwm220fjh, 7qwm220fjh, spinacen
PubChem CID 638072
ChEBI CHEBI:15440
IUPAC Name (6E,10E,14E,18E)-2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene
SMILES CC(C)=CCC\C(C)=C\CC\C(C)=C\CC\C=C(/C)CC\C=C(/C)CCC=C(C)C

Specifications

Melting Point -75°C (literature)
Density 0.858g/mL at 25°C (Lit.)
Boiling Point 285°C (25mm (literature))
Refractive Index n20/D 1.494 (Lit.)
Quantity 1 L
Formula Weight 410.7
Percent Purity ≥98%
Physical Form Liquid
Recommended Storage Room Temperature (15–30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.