Learn More
Chemglass Life Sciences Square Bottle, Media, 125mL, Sterile, PETG, Graduated, 38-430 Tamperproof, Bioprocess, Single Use
SDP
Catalog No. 5000980729
Encompass
Gamma sterilized non-pyrogenic media bottles manufactured from USP Class VI (PETG) polyethylene terephthalate copolyester with tamperproof white polypropylene caps. Tamperproof caps provide a visual indication that the bottle has not been opened once sealed. Once sealed the tamperproof ring will separate from the cap giving a visual indication that the bottle has been opened. Pyrogen, DNase, RNase and Particle-Free. Bottles proved reduced permeability to both CO2 and O2. Note: These bottles are not autoclavable.
Catalog No. 50-009-80729 Supplier Chemglass Life Sciences Supplier No. CLS1168T0125
Gamma sterilized non-pyrogenic media bottles manufactured from USP Class VI (PETG) polyethylene terephthalate copolyester with tamperproof white polypropylene caps. Tamperproof caps provide a visual indication that the bottle has not been opened once sealed. Once sealed the tamperproof ring will separate from the cap giving a visual indication that the bottle has been opened. Pyrogen, DNase, RNase and Particle-Free. Bottles proved reduced permeability to both CO2 and O2. Note: These bottles are not autoclavable.