Learn More
Electron Microscopy Sciences Polypropylene Square Bottles 15 oz, 38 mm cap
SDP
Catalog No. 5028384
Encompass_Preferred
Square polypropylene bottles have screw polypropylene closures and are repeatedly autoclavable and resistant to acids, alcohols and bases. Mouths are wider than conventional narrow mouth bottles. The square shape saves shelf space and the translucent color provides visual liquid level. Useful temperature is between 0 degree C and 135 degree C (32 degree and 275 degree F). Molded-in graduations and drip-proof spout for easier measuring and dispensing. Ideal for buffers, cell culture solutions storage.
Size: 38 mm cap
Catalog No. 50-283-84 Supplier Electron Microscopy Sciences Supplier No. 6424516
Square polypropylene bottles have screw polypropylene closures and are repeatedly autoclavable and resistant to acids, alcohols and bases. Mouths are wider than conventional narrow mouth bottles. The square shape saves shelf space and the translucent color provides visual liquid level. Useful temperature is between 0 degree C and 135 degree C (32 degree and 275 degree F). Molded-in graduations and drip-proof spout for easier measuring and dispensing. Ideal for buffers, cell culture solutions storage.
Size: 38 mm cap