Learn More
Electron Microscopy Sciences Square Mesh Cassettes with Lids Packed Separate, Blue
SDP
Catalog No. 5030254
Encompass_Preferred
Biopsy processing cassettes are similar to the slotted cassettes but have a 1 mm square mesh for small biopsies. Made from Acetyl Polymer, which is a special high density polymer that keeps specimens safely submerged in solution, these cassettes are totally resistant to most histological solvents and their slots offer maximum flow through for good fluid exchange and proper drainage.
The lids can be opened and closed as often as necessary and they always relock securely without danger of specimen loss. Large labeling areas are located on three sides of the cassette for your convenience.
Color: Blue
Catalog No. 50-302-54 Supplier Electron Microscopy Sciences Supplier No. 70072B
Biopsy processing cassettes are similar to the slotted cassettes but have a 1 mm square mesh for small biopsies. Made from Acetyl Polymer, which is a special high density polymer that keeps specimens safely submerged in solution, these cassettes are totally resistant to most histological solvents and their slots offer maximum flow through for good fluid exchange and proper drainage.
The lids can be opened and closed as often as necessary and they always relock securely without danger of specimen loss. Large labeling areas are located on three sides of the cassette for your convenience.
Color: Blue