Learn More
Alkali Scientific Square Cover Glass, Cover Slip, for Microscope Slides, 22 X 22mm, No. 1, Thickness 0.13 to 0.17mm, 10 oz 1000/cs
SDP
Supplier:  Alkali Scientific SM2222C
Encompass
ASI™ Square Cover Glass are high quality imported glass, and are non-cytotoxic. They are manufactured in 100,000 grade clean room and resistant to organic solvents and autoclaved by high temperature.
Catalog No. 50-238-9165