Learn More
Alkali Scientific Square Dish, Clear, PS, Sterile, 10/pk, 240/cs
SDP
Catalog No. 502389403
Encompass_Preferred
This Square Dish Clear is a sterile polystyrene (PS) dish with dimensions of 125x125x20mm. It is packaged with 10 dishes per pack and 240 dishes per case, suitable for a wide range of laboratory uses.
Catalog No. 50-238-9403 Supplier Alkali Scientific Supplier No. TD0125
This Square Dish Clear is a sterile polystyrene (PS) dish with dimensions of 125x125x20mm. It is packaged with 10 dishes per pack and 240 dishes per case, suitable for a wide range of laboratory uses.