Learn More
Bellco Glass, Inc. Square Duran Bottle,500mL
SDP
Catalog No. 501945787
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Square storage bottles are space saving in the autoclave or on the shelf. They are chemically resistant and stable for media preparation and storage. The shoulder groove indicates maximum volume for each size. They are supplied with blue polypropylene linerless screw cap and pouring ring with white graduations and a marking spot.
Catalog No. 50-194-5787 Supplier Bellco Glass, Inc. Supplier No. 563740500
Manufactured from Type 1 Class A Borosilicate Glass. Square storage bottles are space saving in the autoclave or on the shelf. They are chemically resistant and stable for media preparation and storage. The shoulder groove indicates maximum volume for each size. They are supplied with blue polypropylene linerless screw cap and pouring ring with white graduations and a marking spot.