Learn More
MSC Made in USA Square Flexible Bellows, Hypalon, 8 in. x 8 in. x 24 in. L, 1 ea
Catalog No. 5000685828
Encompass
These square flexible bellows provide effective protection for machinery components and equipment, preventing the ingress of dust, debris, and contaminants. Engineered for durability, the bellows are constructed from Hypalon material to maintain integrity in demanding industrial environments.
  • Design ensures flexibility and ease of movement during operation
  • Square shape fits corresponding equipment profiles
  • Durable construction suitable for diverse industrial applications
  • Protects sensitive components from environmental elements
Catalog No. 50-006-85828 Supplier MSC Supplier No. 63415954
These square flexible bellows provide effective protection for machinery components and equipment, preventing the ingress of dust, debris, and contaminants. Engineered for durability, the bellows are constructed from Hypalon material to maintain integrity in demanding industrial environments.
  • Design ensures flexibility and ease of movement during operation
  • Square shape fits corresponding equipment profiles
  • Durable construction suitable for diverse industrial applications
  • Protects sensitive components from environmental elements