Learn More
Alkali Scientific ASI™ Square Glass Coverslips, 24X24mm, 100/sleeve, No. 1, 0.13-0.17mm, 10oz, 1000/cs
SDP
Catalog No. 503028287
Encompass_Preferred
ASI™ Square Glass Coverslips are made from high-quality glass that is non-cytotoxic and manufactured in 100,000 grade clean rooms. Coverslips are 24X24mm. ASI™ cover glass is available with a thickness of No. 1 0.13-0.17mm, No. 1.5 0.17-0.19mm, and No. 2 0.19-.23mm. Optically Clear Glass Cover slip for microscope slides Packed in 100/sleeve or 10 packs of 100, 1000/cs Glass Coverslips are ideal for: Confocal Microscopy Confocal Laser Scanning Microscopy (CLSM) Fluorescence Microscopy Green Fluorescence Protein (GFP) Fluorescence Resonance Energy Transfer (FRET) Immunofluoresence Microscopy Phase Contrast Microscopy Polarized Light Microscopy Differential Interference Contrast (DIC) Multi Photon Laser Scanning Microscopy (MPLSM) High Resolution Image Analysis Infrared Imaging Calcium Imaging
Catalog No. 50-302-8287 Supplier Alkali Scientific Supplier No. SM2424C
ASI™ Square Glass Coverslips are made from high-quality glass that is non-cytotoxic and manufactured in 100,000 grade clean rooms. Coverslips are 24X24mm. ASI™ cover glass is available with a thickness of No. 1 0.13-0.17mm, No. 1.5 0.17-0.19mm, and No. 2 0.19-.23mm. Optically Clear Glass Cover slip for microscope slides Packed in 100/sleeve or 10 packs of 100, 1000/cs Glass Coverslips are ideal for: Confocal Microscopy Confocal Laser Scanning Microscopy (CLSM) Fluorescence Microscopy Green Fluorescence Protein (GFP) Fluorescence Resonance Energy Transfer (FRET) Immunofluoresence Microscopy Phase Contrast Microscopy Polarized Light Microscopy Differential Interference Contrast (DIC) Multi Photon Laser Scanning Microscopy (MPLSM) High Resolution Image Analysis Infrared Imaging Calcium Imaging