Learn More
Electron Microscopy Sciences Maxtaform 100 Mesh, Nickel
SDP
Catalog No. 5028905
Encompass_Preferred
Maxtaform grids with clean and smooth edges, firm support, and a large open area. 3 mm diameter, 0.75 mil thickness.
Grid: Nickel
Pitch: 254 µm
Hole: 213 µm
Bar: 41 µm
:
Catalog No. 50-289-05 Supplier Electron Microscopy Sciences Supplier No. M100NI
Maxtaform grids with clean and smooth edges, firm support, and a large open area. 3 mm diameter, 0.75 mil thickness.
Grid: Nickel
Pitch: 254 µm
Hole: 213 µm
Bar: 41 µm
:
Product Title