Learn More
Electron Microscopy Sciences Embra Square 100 mesh Molybdnum, 65% Open Area
SDP
Catalog No. 5028881
Encompass_Preferred
EMBRA electroformed grids combine a high open area with a rigid construction which allows for relatively easy handling. 3.05 mm diameter, 16 micron thick.
Grid: Molybdnum
Open Area: 65%
Catalog No. 50-288-81 Supplier Electron Microscopy Sciences Supplier No. E100MO
EMBRA electroformed grids combine a high open area with a rigid construction which allows for relatively easy handling. 3.05 mm diameter, 16 micron thick.
Grid: Molybdnum
Open Area: 65%