Learn More
Electron Microscopy Sciences Maxtaform 150 mesh, Nickel
SDP
Catalog No. 5028909
Encompass_Preferred
Maxtaform grids with clean and smooth edges, firm support, and a large open area. 3 mm diameter, 0.75 mil thickness.
Grid: Nickel
Pitch: 165 µm
Hole: 131 µm
Bar: 34 µm
:
Catalog No. 50-289-09 Supplier Electron Microscopy Sciences Supplier No. M150NI
Maxtaform grids with clean and smooth edges, firm support, and a large open area. 3 mm diameter, 0.75 mil thickness.
Grid: Nickel
Pitch: 165 µm
Hole: 131 µm
Bar: 34 µm
:
Product Title