Learn More
Electron Microscopy Sciences Embra Square 200 mesh Aluminum, 50% Open Area
SDP
Catalog No. 5028875
Encompass_Preferred
EMBRA electroformed grids combine a high open area with a rigid construction which allows for relatively easy handling. 3.05 mm diameter, 16 micron thick.
Grid: Aluminum
Open Area: 50%
Catalog No. 50-288-75 Supplier Electron Microscopy Sciences Supplier No. E200AL
EMBRA electroformed grids combine a high open area with a rigid construction which allows for relatively easy handling. 3.05 mm diameter, 16 micron thick.
Grid: Aluminum
Open Area: 50%