Learn More
Electron Microscopy Sciences Maxtaform 200 mesh, Cu/Rh
SDP
Catalog No. 5028904
Encompass_Preferred
Maxtaform grids with clean and smooth edges, firm support, and a large open area. 3 mm diameter, 0.75 mil thickness. Copper grids with one surface coated with inert Rhodium. This coating will eliminate tarnishing, give side identification, and reduce the bar thickness.
Grid: Cu/Rh
Pitch: 127 µm
Hole: 103 µm
Bar: 24 µm
:
Catalog No. 50-289-04 Supplier Electron Microscopy Sciences Supplier No. M200CR
Maxtaform grids with clean and smooth edges, firm support, and a large open area. 3 mm diameter, 0.75 mil thickness. Copper grids with one surface coated with inert Rhodium. This coating will eliminate tarnishing, give side identification, and reduce the bar thickness.
Grid: Cu/Rh
Pitch: 127 µm
Hole: 103 µm
Bar: 24 µm
:
Product Title