Learn More
Electron Microscopy Sciences Maxtaform 200 mesh, Nickel
SDP
Catalog No. 5028910
Encompass_Preferred
Maxtaform grids with clean and smooth edges, firm support, and a large open area. 3 mm diameter, 0.75 mil thickness.
Grid: Nickel
Pitch: 127 µm
Hole: 103 µm
Bar: 24 µm
:
Catalog No. 50-289-10 Supplier Electron Microscopy Sciences Supplier No. M200NI
Maxtaform grids with clean and smooth edges, firm support, and a large open area. 3 mm diameter, 0.75 mil thickness.
Grid: Nickel
Pitch: 127 µm
Hole: 103 µm
Bar: 24 µm
:
Product Title