Learn More
Electron Microscopy Sciences Embra Square 200 mesh Titanium, 50% Open Area
SDP
Catalog No. 5028876
Encompass_Preferred
EMBRA electroformed grids combine a high open area with a rigid construction which allows for relatively easy handling. 3.05 mm diameter, 16 micron thick.
Grid: Titanium
Open Area: 50%
Catalog No. 50-288-76 Supplier Electron Microscopy Sciences Supplier No. E200TI
EMBRA electroformed grids combine a high open area with a rigid construction which allows for relatively easy handling. 3.05 mm diameter, 16 micron thick.
Grid: Titanium
Open Area: 50%