Learn More
Electron Microscopy Sciences Embra Square 300 mesh Aluminum, 40% Open Area
SDP
Catalog No. 5028885
Encompass_Preferred
EMBRA electroformed grids combine a high open area with a rigid construction which allows for relatively easy handling. 3.05 mm diameter, 16 micron thick.
Grid: Aluminum
Open Area: 40%
Catalog No. 50-288-85 Supplier Electron Microscopy Sciences Supplier No. E300AL
EMBRA electroformed grids combine a high open area with a rigid construction which allows for relatively easy handling. 3.05 mm diameter, 16 micron thick.
Grid: Aluminum
Open Area: 40%