Learn More
Electron Microscopy Sciences Maxtaform 300 mesh, Nickel
SDP
Catalog No. 5028901
Encompass_Preferred
Maxtaform grids with clean and smooth edges, firm support, and a large open area. 3 mm diameter, 0.75 mil thickness.
Grid: Nickel
Pitch: 84 µm
Hole: 61 µm
Bar: 23 µm
:
Catalog No. 50-289-01 Supplier Electron Microscopy Sciences Supplier No. M300NI
Maxtaform grids with clean and smooth edges, firm support, and a large open area. 3 mm diameter, 0.75 mil thickness.
Grid: Nickel
Pitch: 84 µm
Hole: 61 µm
Bar: 23 µm
:
Product Title