Learn More
Electron Microscopy Sciences Embra Square 300 mesh Molybdnum, 40% Open Area
SDP
Catalog No. 5028884
Encompass_Preferred
EMBRA electroformed grids combine a high open area with a rigid construction which allows for relatively easy handling. 3.05 mm diameter, 16 micron thick.
Grid: Molybdnum
Open Area: 40%
Catalog No. 50-288-84 Supplier Electron Microscopy Sciences Supplier No. E300MO
EMBRA electroformed grids combine a high open area with a rigid construction which allows for relatively easy handling. 3.05 mm diameter, 16 micron thick.
Grid: Molybdnum
Open Area: 40%