Learn More
Electron Microscopy Sciences TEM Square Grid Mat
SDP
Supplier:  Electron Microscopy Sciences 71160
Encompass_Preferred
White silicone rubber mats, with numbered compartments. Good for organizing grids. They will not slide or jump between compartments. Also ideal for staining grids. Easy to pick up grids without damaging forcep tips.
Catalog No. 50-930-873