Learn More
Electron Microscopy Sciences Veco Square Mesh Handle 100HDspec, Copper
SDP
Catalog No. 5028919
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Copper
Hole: 200 µm
Catalog No. 50-289-19 Supplier Electron Microscopy Sciences Supplier No. HD100SCU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Copper
Hole: 200 µm