Learn More
Electron Microscopy Sciences Veco Square Mesh Handle 111HDspec, Copper
SDP
Catalog No. 5028921
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Copper
Hole: 190 µm
Catalog No. 50-289-21 Supplier Electron Microscopy Sciences Supplier No. HD111SCU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Copper
Hole: 190 µm