Learn More
Electron Microscopy Sciences Veco Square Mesh Handle 111HDspec, Nickel
SDP
Catalog No. 5028917
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Nickel
Hole: 190 µm
Catalog No. 50-289-17 Supplier Electron Microscopy Sciences Supplier No. HD111SNI
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Nickel
Hole: 190 µm